Learn More
Greiner Bio-One 384-well Storage Microplates
GREENER_CHOICE
Catalog No. 07001050 Shop All Greiner Bio-One Products
Change view
Click to view available options
Color:
Clear
Catalog No. Color
07-001-050 Clear
1 options
Catalog No. 07-001-050 Supplier Greiner Bio-One Supplier No. 788860906
Add to Cart
Add to Cart
Reduce volume of liquid necessary for acoustic liquid handling with minimal dead volume.
  • Made from cycloolefin
  • DMSO resistant for compound storage
  • Small volume LoBase
TRUSTED_SUSTAINABILITY
Specifications
Format Standard
Lid Without Lid
Description DMSO resistant
Array 16 x 24
Color Clear
Material Cycloolefin Copolymer
No. of Wells 384
Sterility Non-sterile
Well Shape Flat Bottom (F-well)