Learn More
Greiner Bio-One 384-well Storage Microplates
GREENER_CHOICE
Catalog No. 07001039 Shop All Greiner Bio-One Products
Change view
Click to view available options
Color:
Natural
Catalog No. Color
07-001-039 Natural
1 options
Catalog No. 07-001-039 Supplier Greiner Bio-One Supplier No. 781201906
Add to Cart
Add to Cart
Bind negligible amounts of proteins or active substances.
  • Made from polypropylene
  • Sealed using commercially available heat-sealers
  • Deionised
  • Solid flat bottom
  • Packed in antistatic bags
Service Plan
No
Warranty and Services
No
TRUSTED_SUSTAINABILITY
Specifications
Format Standard
Array 16 x 24
Color Natural
Product Type Storage Microplate
Material Polypropylene
No. of Wells 384
Sterility Non-sterile
Surface Treatment Deionized
Well Shape Flat Bottom (F-well)