Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Poly(propylene carbonate)

Catalog No. 389021100G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
38-902-1100G 100 g
38-902-125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 38-902-1100G Supplier Sigma Aldrich Supplier No. 389021100G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Poly(propylene carbonate) (PPC) is an aliphatic polycarbonate which is mainly used as a packing material and binder resin. The ester group allows the backbone to have molecular flexibility. It is prepared by reacting carbon dioxide and propylene oxide. It has a low thermal stability which can be improved by capping the polymer with active agents.PPC can be used in the electrolytic composites which can be used in the development of electrochemical devices. It is a biocompatible and biodegradable polymer that can be used as a scaffold for potential applications in tissue engineering.

Specifications

CAS 25511-85-7
Linear Formula [CH(CH3)CH2OCO2]n
Quantity 100 g
MDL Number MFCD00197919
Molecular Weight (g/mol) 50000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.