Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Beta-Cyclodextrin, Sulfated Sodium Salt

Catalog No. 3891535G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
38-915-35G 5 g
38-915-325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 38-915-35G Supplier Sigma Aldrich Supplier No. 3891535G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Analytic reagent for enantioselective capillary electrophoresis assays for propafenone and its metabolitesInvestigations into the effect on fluorescence of aminonaphthalimide dyesReactant or reagent for: Complexation of Ru(II) trisdiimine complexesStereoselective sodium borohydride reduction of cyclohexanone derivatives

Specifications

CAS 37191-69-8
Quantity 5 g
MDL Number MFCD00197857
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.