Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich ™-(+)-1-Octyn-3-Ol

Catalog No. 3997011G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
39-970-11G 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Warning
Catalog No. 39-970-11G Supplier Sigma Aldrich Supplier No. 3997011G
Add to Cart
Add to Cart
Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Key intermediate for the synthesis of prostaglandins.

Specifications

CAS 36140-19-9
Density 0.97 g/mL at 25°C
Boiling Point 100°C to 101°C (1 mmHg, lit.)
Molecular Formula C12H22BCl
Refractive Index n20/D 1.442
Linear Formula (C6H11)2BCl
MDL Number MFCD00191906
Quantity 25 mL
Molecular Weight (g/mol) 212.57 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.