Learn More
Medchemexpress LLC HY-U00434 5mg Medchemexpress, 3BDO CAS:890405-51-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYU004345MG
Encompass_Preferred
Medchemexpress, HY-U00434 5mg 3BDO CAS:890405-51-3 3BDO is a new mTOR activator which can also inhibit autophagy . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4446
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.