Learn More
Echelon Biosciences Research Labs 3C-PTDINS(3,4,5)P3 DIC16 100 ug
SDP
Supplier:  Echelon Biosciences Research Labs P3C916100UG
Encompass
PtdIns(3,4,5)P3 3-methylenephosphonate, diC16 (3C-PtdIns(3,4,5)P3, diC16) is a synthetic PI(3,4,5)P3 analog in which the 3-phosphate bond has been rendered metabolically stable as a methylenephosphonate. P-3C908 increases sodium transport across monolayers of cells A6 through activation of GLUT4. This item is non returnable
Catalog No. 50-720-605
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.