Learn More
Corning™ 3D Clear Antibody Buffer
Catalog No. p-7193710 Shop All Corning Products
Change view
Click to view available options
Quantity:
100 mL
Catalog No. Quantity
10-320-108 100 mL
1 options
This item is not returnable. View return policy
MTO (Made-To-Order) item. This item is subject to the following cancellation policy: This item may be cancelled within 10 days of order placement otherwise is not returnable for credit
Catalog No. 10-320-108 Supplier Corning™ Supplier No. 5735
This item is not returnable. View return policy
MTO (Made-To-Order) item. This item is subject to the following cancellation policy: This item may be cancelled within 10 days of order placement otherwise is not returnable for credit
Corning™'s 3D Clear Antibody Buffer is used with immunolabeling in the Corning™ 3D tissue labeling approach. This solution is comprised of PBS/0.2% Tween 20/Heparin/3% donkey serum/5% DMSO.
  • Rapid tissue clearing
  • Easy to Use
  • No Special Equipment Required
  • Compatible with IF, FP and other Fluorescent labels
  • Microplate and Automation Compatible
  • Reversible to Follow up with 2D H&E/IHC
TRUSTED_SUSTAINABILITY
Specifications
Quantity 100 mL
Includes PBS/0.2% Tween 20/Heparin/3% Donkey Serum/5% DMSO
WARNING: Cancer - www.P65Warnings.ca.gov