Learn More
Corning™ 3D Clear Blocking Buffer
Catalog No. 10320110 Shop All Corning Products
Change view
Click to view available options
Quantity:
100 mL
Catalog No. Quantity
10-320-110 100 mL
1 options
MTO (Made-To-Order) item. This item is subject to the following cancellation policy: This item may be cancelled within 10 days of order placement otherwise is not returnable for credit
Catalog No. 10-320-110 Supplier Corning™ Supplier No. 5737
MTO (Made-To-Order) item. This item is subject to the following cancellation policy: This item may be cancelled within 10 days of order placement otherwise is not returnable for credit
Corning™'s 3D Clear Blocking Buffer is used during the Corning™ 3D Clear immunolabeling process. This solution is comprised of PBS/0.2% Triton™ X-100 /6% donkey serum/10% DMSO.
  • Rapid tissue clearing
  • Easy to Use
  • No Special Equipment Required
  • Compatible with IF, FP and other Fluorescent labels
  • Microplate and Automation Compatible
  • Reversible to Follow up with 2D H&E/IHC
TRUSTED_SUSTAINABILITY
Specifications
Quantity 100 mL
Includes PBS/0.2% Triton™ X-100 /6% Donkey Serum/10% DMSO
WARNING: Cancer - www.P65Warnings.ca.gov