Learn More
Corning™ 3D Clear Starter Kit
Catalog No. 10320103 Shop All Corning Products
Change view
Click to view available options
Quantity:
1 Kit
Catalog No. Quantity
10-320-103 1 Kit
1 options
This item is not returnable. View return policy
Catalog No. 10-320-103 Supplier Corning™ Supplier No. 5730
Get Up to $2,500 in Products with Purchase
View all promotions
Add to Cart
Add to Cart
This item is not returnable. View return policy
Corning™'s 3D Clear Starter Kit combines 30 mL of 3D Clear with the four buffer solutions required to process 3D tissue cultures for immunolabeling. Only immunolabels and nuclear stains need to be added. This kit comes with enough material to process three 96-well plates of 3D cell culture models.
  • Rapid tissue clearing
  • Easy to Use
  • No Special Equipment Required
  • Compatible with IF, FP and other Fluorescent labels
  • Microplate and Automation Compatible
  • Reversible to Follow up with 2D H&E/IHC
TRUSTED_SUSTAINABILITY
Specifications
Quantity 1 Kit
Includes 3 D Clear Reagent, Antibody, Blocking, Penetration, and Washing Buffers
WARNING: Cancer - www.P65Warnings.ca.gov