Learn More
Corning™ 3D Clear Tissue Clearing Reagent
Catalog No. 10320105 Shop All Corning Products
Change view
Click to view available options
Quantity:
10 mL
30 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
10-320-105 30 mL
10-320-104 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 10-320-105 Supplier Corning™ Supplier No. 5732
Get Up to $2,500 in Products with Purchase
View all promotions
Add to Cart
Add to Cart
Corning™'s 3D Clear tissue clearing technique is designed specifically to support imaging for 3D cell culture models and plate-based high-throughput processing. When paired with fluorescent labeling (e.g. fluorescent protein, immunofluorescence, chemical dyes) and high content confocal microscopy, Corning™'s 3D Clear Reagent allows for complete 3D cell culture model characterization and more accurate drug screening.
  • Rapid tissue clearing
  • Easy to Use
  • No Special Equipment Required
  • Compatible with IF, FP and other Fluorescent labels
  • Microplate and Automation Compatible
  • Reversible to Follow up with 2D H&E/IHC
TRUSTED_SUSTAINABILITY
Specifications
Quantity 30 mL