Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Corning™ 3D Clear Washing Buffer 10X

Catalog No. p-7193708 Shop All Corning Products
Change view
Click to view available options
Quantity:
200 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
10-320-114 200 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
MTO (Made-To-Order) item. This item is subject to the following cancellation policy: This item may be cancelled within 10 days of order placement otherwise is not returnable for credit
Catalog No. 10-320-114 Supplier Corning™ Supplier No. 5741
Only null left
Add to Cart
Add to Cart
MTO (Made-To-Order) item. This item is subject to the following cancellation policy: This item may be cancelled within 10 days of order placement otherwise is not returnable for credit

Corning™'s 3D Clear Washing Buffer 10X is used for the Corning™ 3D Clear immunolabeling process. This solution is a concentrate and is comprised of: PPBS with 0.2% Tween-20 and 10 μg/mL heparin. This solution should be diluted to 1X concentration with PBS prior to use.

  • Rapid tissue clearing
  • Easy to Use
  • No Special Equipment Required
  • Compatible with IF, FP and other Fluorescent labels
  • Microplate and Automation Compatible
  • Reversible to Follow up with 2D H&E/IHC
TRUSTED_SUSTAINABILITY

Specifications

Quantity 200 mL
Includes PPBS with 0.2% Tween-20 and 10 μg/mL Heparin
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.