Learn More
ILEX LIFE SCIENCES LLC Cellendes 3-D Life CD-Link, Hydrogel Component (3x 200 ul)
SDP
Supplier:  ILEX LIFE SCIENCES LLC L603
Encompass_Preferred
3-D Life is the most flexible system for biomimetic hydrogel design for 3D cell cultures. It uses two polymers that form a hydrogel by covalent crosslinking at ambient temperature. The chemical reaction is non-toxic to cells and occurs rapidly, preventing cell settlement before gel is formed. The CD-Link component is a polyethylene glycol-peptide conjugate with a thiol at each end. Thiol groups on CD-Link form stable thioether bonds with thiol-reactive groups on 3-D Life polymers, resulting in formation of the gel. CD-Link can be used with 3-D Life polymers Mal-PVA, Mal-Dextran, SG-PVA and SG-Dextran. CD-Link contains a matrix metalloproteinase (MMP)-cleavable peptide sequence, allowing cells to locally degrade the polymer network, if they produce the indicated MMPs. Quantity: Cat. No. L60-3: When used with polymers of the 3-D Life Hydrogel system up to 6 ml 3-D Life Hydrogel can be generated, depending on the stiffness of the gel. Manufactured in Germany by Cellendes GmbH.
Catalog No. NC2081823
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.