Learn More
ILEX LIFE SCIENCES LLC Cellendes 3-D Life PVA-HA Hydrogel Ready-To-Design Kit (1 kit)
SDP
Supplier:  ILEX LIFE SCIENCES LLC G851
Encompass_Preferred
3-D Life is the most flexible system for biomimetic hydrogel design for 3D cell cultures. It uses two polymers that form a hydrogel by covalent crosslinking at ambient temperature. The chemical reaction is non-toxic to cells and occurs rapidly, preventing cell settlement before gel is formed. The 3-D Life PVA-HA Hydrogel Kit provides reagents to set up hydrogels containing hyaluronic acid (hyaluronan). Components are mixed at physiological pH 7.2 for optimal cell compatibility. Slow gelation allows for manipulation before onset of gel formation. Cell adhesion peptides (e.g. 3-D Life RGD Peptide, Cat. No. 09-P-001) can be covalently attached to a portion of the SH-reactive groups on SG-PVA to provide a cell-adhesive matrix. One kit allows formation of 3.3 ml hydrogel of a crosslinking strength of 1.2 mmol/L. Manufactured in Germany by Cellendes GmbH.
Catalog No. NC2081801
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.