Learn More
Thermo Scientific™ 3D Tank Liner, Aegis™ 5-14 film, top and bottom drain
Description
- Minimizes bioburden due to construction under sterile cGMP conditions
- Helps eliminate post-use cleaning steps required with reusable containers, reducing cross-contamination risks and reduce cycle times
- Facilitates hydration of powdered media and buffers, and pooling of non-sterile solutions and fluids
Specifications
Specifications
| Capacity (Metric) | 100 L |
| Quantity | 100 L |
| Tubing Style | C-Flex |