Learn More
Epredia™ 3DHistech Pannoramic 250 Flash III Digital Scanner, Brightfield to 80x Resolution, Fluorescence to 90x Resolution - SPECTRA
Catalog No. F3B90SPCS Shop All Epredia Products
Change view
Click to view available options
Resolution:
Brightfield to 80x
Catalog No. Resolution
F3B90SPCS Brightfield to 80x
1 options
Catalog No. F3B90SPCS Supplier Epredia™ Supplier No. F3BF80FL90SPCS
Add to Cart
Add to Cart
Request a Quote
Epredia™ 3DHistech Pannoramic 250 Flash III whole-slide digital scanner is an all-in-one solution for labs that demand BF and/or FL scanning on the same platform with award winning quality and continuous loading of up to 300 slides at FLASH technology speeds.
  • 300-slide capacity and continuous loading with vertical slide arrangement
  • Award-winning, exceptional image quality for both brightfield and up to nine fluorescent filter positions available for single and multiple band cubes with advanced FISH scanning technique
  • Pulsed Xenon FLASH light source for high-speed brightfield scanning and Lumencor SPECTRA light source for fluorescent scanning
  • Up to 80x brightfield and 90x fluorescent resolution by default
  • Darkfield preview for easy localization of fluorescent samples
  • Motorized objective and camera changer
  • Automatic slide loading, previewing, barcode reading and scanning
  • All-around system for high volume slide scanning - Research use only
Specifications
Resolution Brightfield to 80x
Product Type Digital Scanner
For Use With (Application) Digital Pathology
For research and education use only, not for use in diagnostic procedures in the U.S. This product has not been approved or cleared as a medical device by the U.S. Food and Drug Administration.