Learn More
Strem, An Ascensus Company CAS# 15112-89-7. 25g. Tris(dimethylamino)silane, 99+%, 3DMAS (99.999%-Si) PURATREM, 14-8750, contained in 50 ml Swagelok cylinder (96-1070) for CVD/ALD. MFCD00048006
SDP
Supplier:  Strem, An Ascensus Company 98403525G
Encompass_Preferred
CAS# 15112-89-7. 25g. Tris(dimethylamino)silane, 99+%, 3DMAS (99.999%-Si) PURATREM, 14-8750, contained in 50 ml Swagelok cylinder (96-1070) for CVD/ALD. MFCD00048006. Molecular Weight: 161.32. Molecular Formula: [(CH3)2N]3SiH. Color/form: colorless to light yellow liq. Strem# 98-4035. http://www.strem.com/catalog/v/98-4035/
Catalog No. 50-204-5451
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.