Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3F8, Tocris Bioscience™

Catalog No. 408350
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
40-835-0 50 mg
40-831-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 40-835-0 Supplier R&D Systems Supplier No. 4083/50

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This item is not returnable. View return policy

Potent and selective GSK-3β inhibitor

Potent and selective GSK-3β inhibitor (IC50 values are 34 and 304 nM in the presence of 10 and 100 μM ATP respectively).

Chemical Identifiers

CAS 159109-11-2
Molecular Formula C15H14N2O4
Molecular Weight (g/mol) 286.287
InChI Key ULVWJFBHQIXEPE-UHFFFAOYSA-N
Synonym kinome_3660, 5-ethyl-7,8-dimethoxy-1h-pyrrolo 3,4-c isoquinoline-1,3 2h-dione
PubChem CID 23829003
IUPAC Name 5-ethyl-7,8-dimethoxypyrrolo[3,4-c]isoquinoline-1,3-dione
SMILES CCC1=NC2=C(C3=CC(=C(C=C31)OC)OC)C(=O)NC2=O

Specifications

Quantity 50 mg
Formula Weight 286.28
Percent Purity >99%
Chemical Name or Material 3F8
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.