Learn More
3M™ N95 Healthcare Particulate Respirators and Surgical Masks, 1860 Series
Catalog No. 18992 Shop All 3M Products
Change view
Click to view available options
Size:
Small
Standard
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
18-992 Standard
18-991 Small
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18-992 Supplier 3M™ Supplier No. 7000002005
Add to Cart
Add to Cart
Disposable respirators provide protection against certain airborne biological particles. Fluid resistant to splash and spatter of blood and other infectious material.
  • Minimizes wearer exposure to airborne particles generated by electrocautery, laser surgery and powered medical instruments
  • Fluid resistant to splash and spatter of blood and other infectious materials
  • >99% BFE (Bacterial Filtration Efficiency) according to ASTM™ F2101
  • Two sizes available to fit a broad range of faces
  • Contains no components made from natural rubber latex
  • Designed for ease of breathing
  • Braided headbands, cushioning nose foam
TRUSTED_SUSTAINABILITY
Specifications
Aerosol Type N - Not Resistant to Oil
Assigned Protection Factor (APF) 10
Certifications/Compliance ASTM F2101, ASTM F1862
Certifications (CSA) Health Canada Class I Medical Device
Certifications (NIOSH) NIOSH 42 CFR 84, N95, approval number: TC-84A-0006
Certifications (OSHA) OSHA 1910.134
Design Type Cup/Molded
Disposable Single-use
Filter Type Surgical N95
Material Polypropylene, Polyester
Product Type Healthcare Particulate Respirator and Surgical Mask
Size Standard
With Exhalation Valve No
Filter Efficiency 99%
Latex Free Yes
Material Headstrap Braided Polyisoprene
Model 1860
Nose Piece Material Aluminum
Nosepiece Type Adjustable Nose Clip
Resistant To Fluid 120 mmHg
Sterility Non-sterile
Strap Attachment Type Braided and Stapled Headbands
FDA Cleared Surgical Mask Yes