Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ Protective Coverall 4520, Hooded

Catalog No. 17623343 Shop All 3M Products
Change view
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-623-343 X-Large
17-623-341 Medium
17-623-342 Large
17-623-344 2X-Large
17-623-346 4X-Large
17-623-345 3X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 17-623-343 Supplier 3M™ Supplier No. 7000034763
Add to Cart
Add to Cart
This item is not returnable. View return policy

Coveralls offer multiple layers of SMMMS for effective hold out and strength at a lightweight 43 gsm. Breathable and soft with a cloth like feel to be worn for extended periods to protect against static discharge, hazardous particulates, and liquid splash Type 5/6.

  • Type 5/6 Coverall made from extremely lightweight and breathable material, designed to offer protection against hazardous dusts and limited liquid splashes and sprays
  • Made from super breathable SMMMS non-woven for effective release of body heat and moisture and reduced heat stress
  • Soft and strong multi-layered 43 gsm SMS for strong performance
  • 3 panel hood, 2-way zip and storm flap with elasticated back and ankles to reduce the risk of skin exposure
  • Anti-static coating EN 1149-5 reduces static discharge and the risk of sparks
  • EN 1073-2 nuclear particulate hold out
  • Knitted cuffs for soft and secure arm placement
TRUSTED_SUSTAINABILITY

Specifications

Product Type General Purpose Coveralls
Size X-Large
Gender Unisex
Material Polypropylene
Color White
Disposable Single-use
Waist Style Elastic
Ankle Style Elastic
Cuff Style Elastic, Knit
Neck Style Hooded
Hood Type 3-panel
Closure Type Front Zipper with Storm Flap
Seam Type Bound
Sterility Non-sterile
Resistance Dust, Liquid
Coating Anti-static
Certifications/Compliance CE, EN 13034, EN-ISO 13982-1, EN 1149-1:2006
Certifications/Compliance (Europe) CE, EN 13034, EN-ISO 13982-1, EN 1149-1:2006, EN 1073
Description 3 Layers of Melt-blown Polypropylene in-between 2 layers of Spunbound Polypropylene
Latex Free No
Model 4520
Size (English) Chest 43 to 45 in.
Size (English) Inseam 33 in.
Size (English) Waist 52 in.
Size (Metric) Chest 109.22 to 114.3 cm
Size (Metric) Inseam 83.82 cm
Size (Metric) Waist 132.08 cm
For Use With (Application) Insulation Installation, Asbestos and Lead Abatement, Coal Dust in Power Plants, Woodworking, Metal Polishing, Light-Duty Building Cleaning, General Powder Handling, General Industrial Clean-up and Maintenance, Machine or Vehicle Maintenance and Food Processing
Height (English) 5.83 to 6.16 ft.
Height (Metric) 179 to 187 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.