Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ Protective Coverall 4540+, Hooded

Catalog No. 17623358 Shop All 3M Products
Change view
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-623-358 4X-Large
17-623-353 Medium
17-623-354 Large
17-623-355 X-Large
17-623-356 2X-Large
17-623-357 3X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 17-623-358 Supplier 3M™ Supplier No. 7100041420
This item is not returnable. View return policy

Coverall is made from a laminated premium bicomponent non-woven and features a breathable SMMMS triangular back panel for improved comfort. Soft and lightweight material provides an ideal barrier to hazardous particles and limited liquid splashes.

  • Ideal barrier against dry particles and certain liquid chemical splash (CE Type 5/6)
  • Soft and light, ultra low-linting laminated microporous material that drapes and hangs well over the body facilitating ease of movement
  • 3 panel hood, 2-way zip and storm flap with sealable tape with elasticated back and ankles to reduce the risk of skin exposure
  • Knitted cuffs and elasticated waist and ankles for increased comfort and freedom of movement
  • Anti-static coating EN 1149-5 reduces static discharge and the risk of sparks
  • EN 1073-2 nuclear particulate hold out
TRUSTED_SUSTAINABILITY

Specifications

Product Type Protective Coveralls
Size 4X-Large
Gender Unisex
Material Polyethylene, Polyester/Polyethylene Laminate
Color White
Disposable Single-use
Waist Style Elastic
Ankle Style Elastic
Cuff Style Knit
Neck Style Hooded
Hood Type 3-panel
Closure Type Front Zipper with Storm Flap
Seam Type Serged, Bound
Sterility Non-sterile
Protection Class CE Category III, Type 5/6 protection
Resistance Anti-static, Dust, Liquid
Coating Anti-static
Back Style Breathable SMMMS material
Certifications/Compliance (Europe) CE Category III Type 5/6, EN 13034, EN-ISO 13982-1, EN 1149-1:2006, EN 1073
Description White Polypropylene/Polyethylene laminated suit, SMMMS Polypropylene blue back panel, Nylon on Polyester braid zipper, Neoprene Rubber elastic, Polyester cuffs and thread
Latex Free No
Model 4540+
Size (English) Chest 52 to 55 in.
Size (English) Inseam 36 in.
Size (English) Waist 55 in.
Size (Metric) Chest 132.08 to 139.7 cm
Size (Metric) Inseam 91.44 cm
Size (Metric) Waist 139.7 cm
For Use With (Application) Pesticide application, asbestos and lead abatement, Coal dust in power plants, metal polishing, light-duty building Cleaning, machine or vehicle maintenance, paint spraying, pharmaceutical, general industrial clean-up, insulation laying, general powder handling and food processing
Height (English) 6.5 to 6.75 ft.
Height (Metric) 200 to 206 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.