Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ Hooded Anti-Static Protective Coveralls 4565 Series

Catalog No. 17200048 Shop All 3M Products
Change view
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-200-048 2X-Large
17-200-049 Medium
17-623-360 Large
17-623-361 X-Large
17-623-363 3X-Large
17-623-364 4X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 17-200-048 Supplier 3M™ Supplier No. 7100256140
Add to Cart
Add to Cart

Comfortable coveralls made with high-performance fabric offer advanced quality and protection in challenging conditions such as exposure to hazardous dusts and particulates.

  • Type 4/5/6 Coverall designed to offer a very good barrier to hazardous particles and limited liquid splash and spray
  • Made of bicomponent nonwoven with ultra-low linting PE laminate for a noticeably comfortable drape
  • 3 panel hood, 2-way zip and storm flap with sealable tape with elasticated back and ankles to reduce the risk of skin exposure
  • Taped seams across the suit for increased hold out and strength reducing the risk of tearing and exposure
  • Protection against EN 14126 Biological agents
  • Anti-static coating EN 1149-5 reduces static discharge and the risk of sparks
  • EN 1073-2 nuclear particulate hold out Til class 2
TRUSTED_SUSTAINABILITY

Specifications

Product Type Cleanroom Coveralls
Size 2X-Large
Material Microporous Polyethylene Laminate
Color White
Disposable Single-use
Ankle Style Elastic
Cuff Style Knit
Closure Type 2-way Zipper with Sealed Storm Flap
Seam Type Taped
Sterility Non-sterile
Coating Anti-static
Certifications/Compliance ASTM F1670, EN 14126:2003, EN 1149-1:2006, EN 13034, EN-ISO 13982-30
Description Polyethylene and Polyester/Polyethylene laminate suit, Metal/Nylon/Polyester Braid zipper, Neoprene Rubber elastic, Polyester/Cotton thread, Polyester cuffs, Polypropylene red tape
Size (English) Chest 45 to 49 in.
Size (English) Inseam 34 in.
Size (English) Waist 53 in.
Size (Metric) Chest 114.3 to 124.46 cm
Size (Metric) Inseam 86.36 cm
Size (Metric) Waist 134.62 cm
For Use With (Application) Paint Spraying, Industrial and Pharmaceutical Manufacturing, Asbestos Inspection and Handling, Healthcare
Height (English) 6 to 6.33 ft.
Height (Metric) 186 to 194 cm
Cleanroom Class Class 10 (ISO 4)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.