Learn More
Uline 3M 471 Vinyl Tape - 12 x 36 yds, Yellow
Supplier:  Uline S10251Y
Encompass_Preferred
Do-it-all tape color codes masks seals splices and marks hazardous or restricted areas. Conforms to curved and irregular surfaces. Maintains color even with heavy abrasion. Handles temperatures from 40F to 170F. Aggressive rubber adhesive Residue-free removal. 14 LB TENSILE STRENGTH. OSHA Safety Color Compliant.
Catalog No. NC1440899
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.