Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ 6000 Series Resuable Half-Facepiece Respirators with Replaceable P100 Filters

Catalog No. 19003456A Shop All 3M Products
Change view
Click to view available options
For Use With (Application):
Assembly, cleaning, demolition, facility maintenance, grinding, machine operation, painting, sanding, welding
Assembly, cleaning, demolition, facility maintenance, machine operation, painting, sanding
Assembly, cleaning, grinding, machine operation, painting, welding
Format:
Small
Medium
Large
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Application) Format
19-003-456A Assembly, cleaning, demolition, facility maintenance, machine operation, painting, sanding Small
19-003-456B Assembly, cleaning, grinding, machine operation, painting, welding Medium
19-003-456C Assembly, cleaning, demolition, facility maintenance, grinding, machine operation, painting, sanding, welding Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 19-003-456A Supplier 3M™ Supplier No. 7010383694

Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.

This item is not returnable. View return policy

Lightweight, low-maintenance respirators come equipped with replaceable P100 filters (Style 2091)

  • Respirators can be disassembled, cleaned and reused; simply twist off and dispose of filters when they are used to capacity, and attach new filters
  • Effective for dusts, fumes, mists, radionuclides, radon daughters, and asbestos-containing dusts and mists
  • 3M Catalog# 6191 , 3M Catalog# 6291 , 3M Catalog# 6391

These safety products must be used in accordance with OSHA regulations and the user instructions, warnings and limitations accompanying each product. Misuse may result in sickness or death.

Certifications

NIOSH 42 CFR 84 approved for use in areas where oil- and non-oil-based particles are present

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance NIOSH 42CFR84
Connection Type Bayonet
Color Magenta
Material Thermoplastic Elastomer
Product Type Half Facepiece Respirator
Format Small
For Use With (Application) Assembly, cleaning, demolition, facility maintenance, machine operation, painting, sanding
For Use With (Equipment) 6000 series reusable half facepiece respirator
Includes P100 filter
Model 6191
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.