Learn More
3M™ Accessories and Replacement Parts for Full-Facepiece Respirators: Spectacle Kit
Catalog No. 189994558 Shop All 3M Products
Change view
Click to view available options
Spectacle Kit
Catalog No.
18-999-4558 Spectacle Kit
1 options
This item is not returnable. View return policy
Catalog No. 18-999-4558 Supplier 3M™ Supplier No. 7000002031/DEL
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
This item is not returnable. View return policy
Accessories for full-facepiece respirators
  • For full facepiece 6000 Series respirators
  • Permits wearer to use his/her corrective lenses and to make most comfortable position adjustment
  • 3M Catalog# 6878
TRUSTED_SUSTAINABILITY
Specifications
Product Type Spectacle Kit
For Use With (Application) Permits wearer to use his/her corrective lenses and to make most comfortable position adjustment
For Use With (Equipment) 6000 series full facepiece respirator
Model 6878