Learn More
3M™ CO2 Monitor Accessory, Adapter
Catalog No. 19810261 Shop All 3M Products
Change view
Click to view available options
Electrical Requirements:
110/120 V
Catalog No. Electrical Requirements
19-810-261 110/120 V
1 options
Catalog No. 19-810-261 Supplier 3M™ Supplier No. 7000131705
Add to Cart
Add to Cart
For use with compressed air filter and regulator panel and Retrofit CO Monitor
  • Allows convertion of CO from battery operation to AC operation
  • Electrical requirement: 110-120VAC
  • 3M Catalog# 529-04-50
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) CO2 Monitor
Model 529-04-50
Electrical Requirements 110/120 V
Product Type Adapter