Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ Breathe Easy™ Filters, Cartridges, and Canisters

Catalog No. 17987272E Shop All 3M Products
Change view
Click to view available options
Cartridge Type:
Ammonia, Methylamine Cartridge
Filter Cartridge
Organic Vapor Cartridge
Organic Vapor, Chlorine, Hydrogen Chloride, Sulfur Dioxide Cartridge
Organic Vapor, Hydrogen Fluoride, Sulfur Dioxide Cartridge
Color:
Black
Magenta
Magenta/Green
Magenta/Yellow
Product Type:
Combination Cartridge/Filter
HE Filter
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Cartridge Type Color Product Type
17-987-272E Filter Cartridge Magenta HE Filter
17-987-272F Organic Vapor Cartridge Black Combination Cartridge/Filter
17-987-272G Organic Vapor, Chlorine, Hydrogen Chloride, Sulfur Dioxide Cartridge Black Combination Cartridge/Filter
17-987-272J Ammonia, Methylamine Cartridge Magenta/Green Combination Cartridge/Filter
17-988-398 Organic Vapor, Hydrogen Fluoride, Sulfur Dioxide Cartridge Magenta/Yellow Combination Cartridge/Filter
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 17-987-272E Supplier 3M™ Supplier No. 7000131700
This item is not returnable. View return policy

Used for protection against a combination of organic vapors, hydrogen fluoride, sulfur dioxide and/or high efficiency particulates.

  • NIOSH-approved P3 particulate cartridges used for Breathe Easy PAPR systems
  • High efficiency (HE) respiratory protection against particulates
  • 3M Catalog# 450-00-01R12, 453-00-01R06, 453-02-01R06, 453-03-01R06, 453-07-01R06
TRUSTED_SUSTAINABILITY

Specifications

Cartridge Type Filter Cartridge
Certifications/Compliance NIOSH, MSHA
Color Magenta
Filter Type High Efficiency (HE)
Product Type HE Filter
For Use With (Equipment) Breathe Easy™ PAPR Systems
Model 450-00-01R12
Quantity 12/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.