Learn More
Description
- NIOSH-approved P3 particulate cartridges used for Breathe Easy PAPR systems
- High efficiency (HE) respiratory protection against particulates
- 3M Catalog# 450-00-01R12, 453-00-01R06, 453-02-01R06, 453-03-01R06, 453-07-01R06
Specifications
Specifications
| Cartridge Type | Organic Vapor Cartridge |
| Certifications/Compliance | NIOSH, MSHA |
| Color | Black |
| Filter Type | High Efficiency (HE) |
| Product Type | Combination Cartridge/Filter |
| For Use With (Equipment) | Breathe Easy™ PAPR Systems |
| Model | 453-00-01R06 |
| Quantity | 6/Pk. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.