Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
3M™ 3M™ Brown Combat High Cut Ballistic Helmet with CamFit™ Retention System
Manufactured with similar dimensions to the U.S. Military ACH helmet, but with a higher cut. 3M Company™ 3M™ Brown Combat High Cut Ballistic Helmet with CamFit™ Retention System allow for unobstructed views and increased hearing capabilities.
Specifications
| Color | Brown |
|---|---|
| Includes | 7 Pad System and Team Wendy™ Dial Adjustable CamFit™ Retention System |
| Product Type | Ballistic Helmet |
| Quantity | 1 Ea. |
| Catalog Number | Mfr. No. | Size | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Size | Price | Quantity & Availability | |||||
|
03-052-817
|
3M™
98009007174 |
Small | N/A | N/A | N/A | ||||
|
03-051-092
|
3M™
98009007182 |
Medium | N/A | N/A | N/A | ||||
|
03-051-103
|
3M™
98009007190 |
Large | N/A | N/A | N/A | ||||
|
03-052-805
|
3M™
98009007208 |
X-Large | N/A | N/A | N/A | ||||
Description
Specifications
| Brown | |
| 7 Pad System and Team Wendy™ Dial Adjustable CamFit™ Retention System | |
| Ballistic Helmet | |
| 1 Ea. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title