Learn More
3M™ 7800S Series Full Facepiece Respirator Accessory, Cartridge Filter Adapter 701
Replacement part for 7800S Series full-facepiece respirators
Supplier: 3M™ 7000126044
Description
- Use to attach 2000 Series filters, 7093 filter, 6000 Series Cartridges or dual airline system
- Comes fully assembled — simply attach to facepiece and it's ready
- Wearer can then position cartridge or filter for greatest comfort and maximum visibility
- 3M Catalog# 701
These safety products must be used in accordance with OSHA regulations and the user instructions, warnings and limitations accompanying each product. Misuse may result in sickness or death.
Order Info
Each pack contains two adapters.