Learn More
3M™ Combat High Cut Ballistic Helmet with 3pt Night Vision Goggle Shroud
Catalog No. 03051099 Shop All 3M Products
Change view
Click to view available options
Size:
Large
Medium
Small
X-Large
Color:
Black
Green
Tan
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Color
03-051-099 X-Large Green
03-052-810 Small Black
17-000-646 Medium Black
03-052-816 X-Large Black
03-052-809 Small Green
03-052-811 Medium Green
03-052-806 Large Green
03-051-095 Small Tan
03-051-096 Medium Tan
03-052-803 Large Tan
03-052-802 X-Large Tan
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
Catalog No. 03-051-099 Supplier 3M™ Supplier No. 7010350762
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.
Manufactured with similar dimensions to the U.S. Military ACH helmet. 3M™ Combat High Cut Ballistic Helmet with 3pt Night Vision Goggle Shroud features a higher cut for unobstructed views and increased hearing capabilities, compared to the PASGT helmet.
  • Geometry is similar to the U.S. Army Advanced Combat Helmet (ACH)
  • Available in a Tactical High Cut configuration
  • Designed to accommodate night vision goggle shroud with minimum loss of vision or mobility
  • Includes Night Vision Goggle (NVG) Shroud
  • Manufactured utilizing advanced thermoplastic aramid and glass fabric composites
  • Retention system delivers a stable and secure fit along with a user-friendly quick release for easy on, easy off use when sighting a target
  • 3M Catalog# 98009009352, 98009009428, 98009009394, 98009009337, 98009009386, 98009009444, 98009009410, 98009009345, 98009009360, 98009009436, 98009004130
TRUSTED_SUSTAINABILITY
Specifications
Color Green
Product Type Ballistic Helmet
Quantity 1 Ea.
Size X-Large