Learn More
MEDPLUS SERVICES USA 3M™ Comply™ Bowie-Dick Plus Test Pack
Supplier:  MEDPLUS SERVICES USA MMM 00135LF
Encompass_Preferred
Early warning feature of the 3M™ Comply™ Bowie-Dick Plus Test Pack 00135LF permits scheduled sterilizer maintenance and helps to prevent unexpected failures. Indicator sheet will turn a uniform dark brown/black color except when air removal failures, such as air leaks, occur. Can be filed for record keeping.. Ink is not made with lead materials.
Catalog No. NC1335409
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.