Learn More
MEDPLUS SERVICES USA Steam Chemical Integrator, 2" x 3/4", 1000 count
Supplier:  MEDPLUS SERVICES USA MMM 1243B
Encompass_Preferred
Steam Chemical Integrator,1000/pack, 2 inch x 3/4/inch ,Chemical integrators are used for pack control which validates that the sterilant has penetrated to the point of placement in the pack
Catalog No. NC9813091
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.