Learn More
Description
- Made with proprietary Advanced Electrostatic Media (AEM)
- Particulate filters are both flame and water resistant
- Easy to install
- Particulate Filters 2000 Series are compatible with Half Facepieces 6000, 6500/6500QL and 7500 Series, and Full Facepieces 6000, FF-400 and 7800 Series
Specifications
Specifications
| Cartridge Type | OV |
| Connection Type | Bayonet |
| Color | Pink |
| Filter Type | P |
| Product Type | Filter |
| Filter Efficiency | 100 |
| For Use With (Equipment) | 3M 6000, 6500, 7500, or FF-400 |
| Model | 50570-00000 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.