Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ Disposable Protective Coveralls 4540+, Collared

Catalog No. 17624113 Shop All 3M Products
Change view
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-624-113 X-Large
17-624-110 4X-Large
17-624-111 Large
17-624-109 3X-Large
17-624-112 Medium
17-624-114 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 17-624-113 Supplier 3M™ Supplier No. 7100213267
This item is not returnable. View return policy

Coveralls are designed to help protect against hazardous dusts (Type 5) and certain light liquid splashes (Type 6).

  • Premium dual material technology provides both protective performance and comfort
  • Barrier against dry particles and limited liquid chemical splash (CE Type 5/6)
  • Soft and light, super low-linting laminated microporous material
  • Back panel made of breathable SMS material for improved air circulation and reduced heat stress
  • Two-way zipper with fully sealable storm flap
  • Fold down collared design
  • Knitted cuffs and elasticated waist and ankles for increased comfort and freedom of movement
  • Anti-static treated
  • Low linting
TRUSTED_SUSTAINABILITY

Specifications

Product Type General Purpose Coveralls
Size X-Large
Gender Unisex
Material Polyethylene, Polyester/Polyethylene Laminate
Color White
Disposable Single-use
Waist Style Elastic
Ankle Style Elastic
Cuff Style Knit
Neck Style Collared
Hood Type Unhooded
Closure Type Front Zipper with Storm Flap
Seam Type Serged, Bound
Sterility Non-sterile
Resistance Anti-static, Dust, Liquid
Coating Anti-static
Collar Type Standard Fold
Certifications/Compliance ANSI 101-1996
Certifications/Compliance (Europe) CE Category III Type 5/6, EN 13034, EN-ISO 13982-1, EN 1149-1:2006, EN 1073
Description White Polypropylene/Polyethylene laminated suit, SMMMS Polypropylene blue back panel, Nylon on Polyester braid zipper, Neoprene Rubber elastic, Polyester cuffs and thread
Latex Free No
Model 4540+
Size (English) Chest 43 to 45 in.
Size (English) Inseam 33 in.
Size (English) Waist 52 in.
Size (Metric) Chest 108 to 115 cm
Size (Metric) Waist 132 cm
For Use With (Application) Pesticide, Asbestos and Lead Abatement, Power Plants, Metals, Cleaning, Maintenance, General Industrial, Insulation, Food Processing, Pharmaceutical
Height (English) 70 to 74 in.
Height (Metric) 179 to 187 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.