Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ Disposable Protective Lab Coat, Series 4440

Catalog No. 17624127 Shop All 3M Products
Change view
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-624-127 Medium
17-624-129 X-Large
17-624-130 2X-Large
17-624-132 4X-Large
17-624-128 Large
17-624-131 3X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 17-624-127 Supplier 3M™ Supplier No. 7000089709
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy

Offers a barrier against non-hazardous particles and liquid splashes in light duty applications.

  • Breathable, lightweight material.
  • Designed for light duty applications.
  • Offers a barrier against non-hazardous particles and liquid splashes.
  • Zip fastener.
  • Anti-static treated.
  • Knitted cuffs.
TRUSTED_SUSTAINABILITY

Specifications

Product Type General Purpose Lab Coat
Size Medium
Gender Unisex
Material Polypropylene
Color White
Closure Type Zipper
Cuff Style Knit
Disposable Single-use
Model 4440-M
Seam Type Serged
Size (English) Chest 32 to 36 in.
Size (Metric) Chest 81 to 91 cm
Certifications/Compliance (Europe) CE Category 1
For Use With (Application) Catering, Cleanrooms, Electronics Manufacturing, General Maintenance, Laboratories
Weight (English) 1.93 oz.
Construction Microporous Laminate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.