Description
- Slow recovery, low-pressure foam conforms to the shape of the ear canal
- Moisture-resistant helps maintain a consistent expansion time, even in humid environments
- Flame resistant for potential arc flash situations and for storage considerations
Compliance
3M™ ear plugs must be used in accordance with OSHA regulations, and the user instructions, warnings, and limitations accompanying each product.
Specifications
Specifications
| ANSI S3.1974 | Yes |
| Cord Color | Blue |
| Cord Material | Vinyl |
| CSA Z94.2 | Class AL |
| Size | Regular |
| For Use With (Application) | Assembly, Blasting, Cleaning, Demolition, Electrical, Facility Maintenance, Grinding, Machine Operations, Painting, Sanding, Welding |
| High Visibility | Yes |
| Model | 311-1081 |
| Packaging Type | Econopack dispenser box |
| Pairs per Package | 1500 Pairs/Cs |
| Show More |