Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ E-A-R™ Classic™ Plus and Classic Small Ear Plugs

Catalog No. 189994504 Shop All 3M Products
Change view
Click to view available options
Model:
310-1101
310-1103
311-1105
311-1106
Corded:
Corded
Uncorded
Size:
Small
Large
Packaging Type:
Pillow Pk.
Poly bag
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Model Corded Size Packaging Type
18-999-4504 311-1106 Corded Small Poly bag
18-999-4507 311-1105 Corded Large Poly bag
18-999-4505 310-1103 Uncorded Small Pillow Pk.
18-999-4506 310-1101 Uncorded Large Pillow Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 18-999-4504 Supplier 3M™ Supplier No. 7000127173
This item is not returnable. View return policy

E-A-R™ Classic™ Plus (NRR 33dB) ear plugs are 20% longer than the regular E-A-R Classic ear plug (and the same diameter) to fit deeper into the ear canal and help provide higher noise reduction. ear Classic Small (NRR 29dB) ear plugs are 7% less than that of the Classic ear plug (and the same length) to help provide comfort and protection in smaller ears.

  • Conforms to the shape of the ear canal to help create a comfortable barrier
  • Slow-recovery ear plug foam is flame-resistant and moisture-resistant
  • Test compatible with 3M™ E-A-Rfit™ Validation System
  • Classic Plus ear plugs are slightly longer than regular E-A-R Classic ear plugs to fit deeper into the ear canal and provide higher noise reduction
  • Classic Small ear plugs are smaller in diameter than regular E-A-R Classic ear plugs to increase comfort in medium and small ear canals while retaining excellent noise attenuation

Compliance

3M™ ear plugs must be used in accordance with OSHA regulations, and the user instructions, warnings, and limitations accompanying each product.

TRUSTED_SUSTAINABILITY

Specifications

ANSI S3.1974 Yes
Cord Color Blue
Cord Material Vinyl
CSA Z94.2 Class AL
Size Small
For Use With (Application) Assembly, Blasting, Cleaning, Demolition, Electrical, Facility Maintenance, Grinding, Machine Operations, Painting, Sanding, Welding
High Visibility Yes
Model 311-1106
Packaging Type Poly bag
Pairs per Package 200 Pairs/Pk.
Plug Material Foam
Plug Color Yellow
Noise Reduction Rating (NRR) 33 dB
Corded Corded
Plug Shape Cylinder
Disposable Single-use
Metal Detectable No
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.