Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
3M™ E-A-R™ Express™ Pod Plugs™ Ear Plugs
Express™ Pod Plugs™ ear plugs allows the foam to compress so the ear plug slides gently into the ear and expands slowly. Flexible stem makes inserting easy.
$105.96 - $170.39
Specifications
ANSI S3.1974 | Yes |
---|---|
CSA Z94.2 | Class AL |
Size | Universal |
For Use With (Application) | Assembly, Blasting, Cleaning, Demolition, Electrical, Facility Maintenance, Grinding, Machine Operations, Painting, Sanding, Welding |
High Visibility | Yes |
Catalog Number | Mfr. No. | Model | Corded | Product Type | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Model | Corded | Product Type | Price | Quantity & Availability | |||
18-999-600
|
3M™
7000002309 |
311-1114 | Corded | Blue grip ear plug |
Pack of 100 for $170.39
Case of 4 PK for $681.54
|
|
|||
18-586
|
3M™
7000002308 |
321-2200 | Uncorded | Assorted color grip ear plug |
Pack of 100 for $114.89
Case of 4 PK for $459.55
|
|
|||
18-587
|
3M™
7000127178 |
321-2100 | Uncorded | Blue grip ear plug |
Pack of 100 for $105.96
Case of 4 PK for $423.82
|
|
|||
18-763
|
3M™
7000127179 |
311-1115 | Corded | Assorted color grip ear plug | N/A | N/A | N/A | ||
Description
- Paddle-style grips provide quick, clean, and consistent placement of ear plugs
- No-roll design keeps ear plugs and ears clean, even when fingers or gloves are dirty
- Slow recovery foam easily goes into ear canal and then seats and stays in place
Compliance
3M™ ear plugs must be used in accordance with OSHA regulations, and the user instructions, warnings, and limitations accompanying each product.
Specifications
Yes | |
Universal | |
Yes | |
400 Pairs/Pk. | |
Yellow | |
Pod | |
No |
Class AL | |
Assembly, Blasting, Cleaning, Demolition, Electrical, Facility Maintenance, Grinding, Machine Operations, Painting, Sanding, Welding | |
Pillow Pk. | |
Foam, thermoplastic | |
25 dB | |
Reusable |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title