Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ E·A·Rsoft™ Yellow Neons™ Ear Plugs

NRR: 33dB

Manufacturer:  3M™ 3111250

 View more versions of this product

Catalog No. 19-041-203



  Check Availability
Add to cart

Alert:

The 3M products in this catalog must be used in accordance with OSHA regulations, and the user instructions, warnings and limitations accompanying each product.


Specifications Description & Specifications

Specifications

9.25 in.
Regular
Vinyl
Yes
E-A-Rsoft™ Yellow Neons™ Earplug
12 in.
30.4cm
Tapered
Corded, Poly Bag; Pk. of 200
33dB
9.25 x 12.7 x 12 in. (23.4 x 15.8 x 30.4cm)
23.4cm
Yellow
Yes
Tapered design allows for easy insertion and removal
Roll down foam
Polyurethane
Aircraft, ships, vehicles
15.8cm
12.7 in.
CSA Class AL
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit