Learn More
3M™ Fuel™ Protective Eyewear
Catalog No. 191303917 Shop All 3M Products
Change view
Click to view available options
Frame Color:
Metallic Sand
Metallic Silver
Lens Tint:
Blue Mirror
Red Mirror
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Frame Color Lens Tint
19-130-3917 Metallic Silver Blue Mirror
19-130-3916 Metallic Sand Red Mirror
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
The 3M products in this catalog must be used in accordance with OSHA regulations, and the user instructions, warnings, and limitations accompanying each product.
Catalog No. 19-130-3917 Supplier 3M™ Supplier No. 7000029997
This item is not returnable. View return policy
The 3M products in this catalog must be used in accordance with OSHA regulations, and the user instructions, warnings, and limitations accompanying each product.
Includes
Microfiber bag and lanyard
Durable nylon frame with polycarbonate lens
  • Scratch-resistant lens coating
  • Lenses absorb 99.9% UV
  • Soft nose bridge and temple tips for added comfort
  • Vented temples
  • Fuel2 Model (sold separately) has larger lenses
  • 3M Catalog# 11641-00000-10 , 3M Catalog# 11640-00000-10
Compliance
Meets ANSI™ Z87.1-2010 standards. Fuel2 also meets CSA Z94.3-2007 standards.
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance Meets ANSI Z87.1-2010 standards. Fuel2 also meets CSA Z94.3-2007 standards.
Lens Tint Blue Mirror
Product Type Wraparound protective eyewear
For Use With (Application) Eye protection builded to withstand the rigors of job sites
Frame Color Metallic Silver
Includes Microfiber bag and lanyard
No. per Case 10/Cs.
Temple Style Vented
Lens Material Polycarbonate
Frame Material Nylon
Frame Type Full frame