Learn More
3M™ GVP-Series PAPR Accessory, Belts
Catalog No. 19169005 Shop All 3M Products
Change view
Click to view available options
Material:
Leather
Vinyl
Web
Product Type:
Belt
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material Product Type
19-169-005 Leather Belt
19-813-773 Vinyl Belt
19-813-775 Web Belt
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
All safety products must be used in accordance with OSHA regulations and with the user instructions, warnings, and limitations accompanying each product. Misuse may result in sickness or death.
Catalog No. 19-169-005 Supplier 3M™ Supplier No. 7000126406
Add to Cart
Add to Cart
This item is not returnable. View return policy
All safety products must be used in accordance with OSHA regulations and with the user instructions, warnings, and limitations accompanying each product. Misuse may result in sickness or death.
For use with GVP-Series powered air purifying respirators.
  • Part of the 3M™ Belt-Mounted Powered Air Purifying Respirator (PAPR) Assembly GVP-1
  • Available in 3 different materials
  • Designed to hold the motor blower unit and battery pack in place
  • 3M Catalog# CB-1000 , 3M Catalog# GVP-117 , 3M Catalog# GVP-127
TRUSTED_SUSTAINABILITY
Specifications
Disposable Reusable
Material Leather
Product Type Belt
For Use With (Application) Assembly, Cleaning, Electrical, Facility Maintenance, Machine Operation, Painting, Sanding, Welding
For Use With (Equipment) GVP Series PAPR
Model CB-1000