Description
- Suspension adjusts easily to allow for customized height and fit
- Replaceable brow pad for more comfort and extended shelf life
- Short brim for upward visibility
- Meets requirements of ANSI/ISEA Z89.1-2009 Type 1, Class C
- ANSI/ISEA compliant when worn in the forward and reverse donning positions
- Available in various colors
- 3M Catalog# H-701R, H-702R, H-703R, H-704R, H-705R, H-706R, H-707R, H-708R, H-709R, H-710R, H-711R, H-712R, H-713R
Specifications
Specifications
| Certifications/Compliance | Meets ANSI/ISEA Z89.1-2009 Type 1, Class C, G and E |
| Color | Bright Orange |
| Includes | Standard brow pad |
| Model | H-707R |
| Product Type | Cap Style Hard Hat |
| Quantity | 20/Cs. |
| Suspension Type | 4-Point Ratchet |