Learn More
3M™ H-700 Series Cap Style Hard Hats with 4-Point Ratchet Suspension
Catalog No. 17623326 Shop All 3M Products
Change view
Click to view available options
Color:
Black
Blue
Bright Orange
Bright Yellow
Gray
Green
Navy Blue
Orange
Pink
Red
Tan
White
Yellow
13 product options available for selection
Product selection table with 13 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
17-623-326 Green
17-623-312 White
17-623-313 Blue
17-623-314 Gray
17-623-321 Navy Blue
17-623-322 Bright Yellow
17-623-323 Bright Orange
17-623-324 Orange
17-623-325 Red
17-623-327 Yellow
17-623-785 Tan
17-623-790 Black
17-623-795 Pink
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 13 options available.
13 options
Catalog No. 17-623-326 Supplier 3M™ Supplier No. 7000030062
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.
Protect top of the head due to potential hazard of small falling objects or exposed fixed objects
  • Suspension adjusts easily to allow for customized height and fit
  • Replaceable brow pad for more comfort and extended shelf life
  • Short brim for upward visibility
  • Meets requirements of ANSI/ISEA Z89.1-2009 Type 1, Class C
  • ANSI/ISEA compliant when worn in the forward and reverse donning positions
  • Available in various colors
  • 3M Catalog# H-701R, H-702R, H-703R, H-704R, H-705R, H-706R, H-707R, H-708R, H-709R, H-710R, H-711R, H-712R, H-713R
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance Meets ANSI/ISEA Z89.1-2009 Type 1, Class C, G and E
Color Green
Includes Standard brow pad
Model H-704R
Product Type Cap-style Hard Hat
Quantity 20/Cs.
Suspension Type 4-Point Ratchet