Learn More
3M™ H-700 Series Cap Style Vented Hard Hats
Catalog No. 17623318 Shop All 3M Products
Change view
Click to view available options
Color:
Black
Blue
Green
Pink
Tan
White
Yellow
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
17-623-318 Blue
17-623-319 Yellow
17-623-320 White
17-623-317 Green
17-623-787 Tan
17-623-792 Black
17-623-797 Pink
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Catalog No. 17-623-318 Supplier 3M™ Supplier No. 7000002420
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.
This item is not returnable. View return policy
Helps protect top of the head due to potential hazard of small falling objects or exposed fixed objects
  • Suspension allows to customize height and fit
  • Vents allow air circulation to keep worker cooler and more comfortable
  • Meets requirements of ANSI/ISEA Z89.1-2009 Type 1, Class C
  • ANSI/ISEA compliant when worn in the forward and reverse donning positions
  • Replaceable brow pad for comfort and extends service life
  • Short brim for upward visibility
  • Available in various colors
  • 3M Catalog# H-701V, H-702V, H-703V, H-704V, H-711V, H-712V, H-713V
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance ANSI/ISEA Z89.1-2014, Type 1
Color Blue
Includes Standard brow pad
Model H-703V
Product Type Cap-style Hard Hat
Quantity 20/Cs.
Suspension Type 4-Point Ratchet
Ventilated Ventilated