Learn More
3M™ H-700 Series Cap Style Vented Hard Hats
Catalog No. 17623320 Shop All 3M Products
Change view
Click to view available options
Color:
Black
Blue
Green
Pink
Tan
White
Yellow
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
17-623-320 White
17-623-318 Blue
17-623-319 Yellow
17-623-317 Green
17-623-787 Tan
17-623-792 Black
17-623-797 Pink
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 17-623-320 Supplier 3M™ Supplier No. 7000002418
Helps protect top of the head due to potential hazard of small falling objects or exposed fixed objects
  • Suspension allows to customize height and fit
  • Vents allow air circulation to keep worker cooler and more comfortable
  • Meets requirements of ANSI/ISEA Z89.1-2009 Type 1, Class C
  • ANSI/ISEA compliant when worn in the forward and reverse donning positions
  • Replaceable brow pad for comfort and extends service life
  • Short brim for upward visibility
  • Available in various colors
  • 3M Catalog# H-701V, H-702V, H-703V, H-704V, H-711V, H-712V, H-713V
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance ANSI/ISEA Z89.1-2014, Type 1
Color White
Includes Standard brow pad
Model H-701V
Product Type Cap Style Hard Hat
Quantity 20/Cs.
Suspension Type 4-Point Ratchet
Ventilated Ventilated