Learn More
3M™ H-800 Series Full Brim Hard Hats
Catalog No. p-4625723 Shop All 3M Products
Change view
Click to view available options
Color:
Black
Blue
Gray
Green
Hi-vis Orange
Hi-vis Yellow
Navy Blue
Orange
Pink
Red
Tan
White
Yellow
13 product options available for selection
Product selection table with 13 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
17-623-706 White
17-623-707 Yellow
17-623-709 Green
17-623-710 Red
17-623-708 Blue
17-623-711 Orange
17-623-712 Hi-vis Orange
17-623-713 Gray
17-623-714 Hi-vis Yellow
17-623-716 Tan
17-623-715 Navy Blue
17-623-772 Black
17-623-776 Pink
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 13 options available.
13 options
Catalog No. 17-623-706 Supplier 3M™ Supplier No. 7000144957
Helps protect the head from small falling objects and fixed objects
  • Made of HDPE
  • Features 4-point ratchet suspension with three position height adjustment allows for customized fit
  • Perforated absorbent brow pad 
  • Optional 6-point suspension strap for more comfort
  • Meets requirements of ANSI/ISEA Z89.1-2009 Type 1, Class C, G and E
  • Available in several colors
  • 3M Catalog# H-801R, H-802R, H-803R, H-804R, H-805R, H-806R, H-807R, H-808R, H-809R, H-810R, H-811R, H-812R, H-813R
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance ANSI/ISEA Z89.1-2009 Type I, Class C, G and E
Color White
Includes 4-point Ratchet Suspension and Standard Brow Pad
Material HDPE
Model H-801R
Quantity 20/Cs.
Size (Metric) 168 to 197 mm
Suspension Type 4-point Ratchet
Weight (English) 416 oz.
Weight (Metric) 11.79 kg
Ventilated Not Ventilated
Head Style Full Brim