Learn More
3M™ H-800 Series Full Brim Hard Hats with Uvicator™ Sensors
Catalog No. 17623734 Shop All 3M Products
Change view
Click to view available options
Color:
Black
Blue
Gray
Green
Hi-vis Orange
Hi-vis Yellow
Navy Blue
Orange
Pink
Red
Tan
White
Yellow
13 product options available for selection
Product selection table with 13 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
17-623-734 Hi-vis Orange
17-623-729 Yellow
17-623-730 Blue
17-623-731 Green
17-623-728 White
17-623-732 Red
17-623-733 Orange
17-623-735 Gray
17-623-737 Navy Blue
17-623-736 Hi-vis Yellow
17-623-738 Tan
17-623-773 Black
17-623-777 Pink
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 13 options available.
13 options
Catalog No. 17-623-734 Supplier 3M™ Supplier No. 7000144981
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.
With Uvicator sensor that changes color as hard hat is exposed to UV light
  • Made of HDPE
  • Features 4-point ratchet suspension with three position height adjustment allows for customized fit
  • Perforated absorbent brow pad 
  • Optional 6-point suspension strap for more comfort
  • Meets requirements of ANSI/ISEA Z89.1-2009 Type 1, Class C, G and E
  • Available in several colors
  • 3M Catalog# H-801R-UV, H-802R-UV, H-803R-UV, H-804R-UV, H-805R-UV, H-806R-UV, H-807R-UV, H-808R-UV, H-809R-UV, H-810R-UV, H-811R-UV, H-812R-UV, H-813R-UV
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance ANSI Z89.1-2009 High-visibility Performance, ANSI/ISEA Z89.1-2009 Type I Class C, G and E
Color Hi-vis Orange
Includes Uvicator™ Sensor, 4-point Ratchet Suspension and Standard Brow Pad
Material HDPE
Model H-807R-UV
Quantity 20/Cs.
Size (Metric) 168 to 197 mm
Suspension Type 4-point Ratchet
Weight (English) 416 oz.
Weight (Metric) 11.79 kg
Ventilated Not Ventilated
Head Style Full Brim