Learn More
3M™ H-800 Series Full Brim Vented Hard Hats
Catalog No. 17623723 Shop All 3M Products
Change view
Click to view available options
Color:
Black
Blue
Gray
Green
Hi-vis Orange
Hi-vis Yellow
Navy Blue
Orange
Pink
Red
Tan
White
Yellow
13 product options available for selection
Product selection table with 13 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
17-623-723 Hi-vis Orange
17-623-718 Yellow
17-623-719 Blue
17-623-717 White
17-623-720 Green
17-623-722 Orange
17-623-721 Red
17-623-724 Gray
17-623-725 Hi-vis Yellow
17-623-727 Tan
17-623-726 Navy Blue
17-623-774 Black
17-623-778 Pink
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 13 options available.
13 options
Catalog No. 17-623-723 Supplier 3M™ Supplier No. 7000144971
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.
3M™ H-800V Series Full Brim Vented Hard Hat with Ratchet System is comfortable, tough and lightweight protective headgear that helps protect the head from small falling objects and fixed objects that may be bumped into. Vents built into the hard hat release heat buildup and allow air circulation to enhance comfort of the worker. The cooling effect of air circulation helps reduce perspiration in hot and humid working environments. The 4-point suspension system has unique features that allow for a customized height and fit that best suits the individual needs of the wearer. An optional 6-point suspension strap can be added for additional comfort and fit. The comfortable suspension sits lower on the head to reduce pressure points and increase security, especially when looking up and bending down. A smooth, easy-turning ratchet suspension allows for simple adjustments. A 3-level height adjustment allows workers to customize the fit. The perforated, absorbent brow pad provides enhanced comfort.
  • Impact resistant, ANSI Type I hard hat wears lightweight and comfortable on the head
  • Comfortable suspension sits lower on head to reduce pressure points
  • Smooth, easy turning ratchet suspension is easy to adjust
  • 4-point ratchet suspension with three position height adjustment allows for customized fit
  • Vents allow air circulation to keep worker cooler and more comfortable
  • Meets requirements of ANSI/ISEA Z89.1-2014 Type 1, Class C
  • High Visibility color meets ANSI Z89.1-2014 High Visibility Performance Requirements.
  • 3M Catalog# H-801V, H-802V, H-803V, H-804V, H-805V, H-806V, H-807V, H-808V, H-809V, H-810V, H-811V, H-812V, H-813V
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance ANSI/ISEA Z89.1-2014, Type 1
Color Hi-vis Orange
Includes 4-point Ratchet Suspension and Standard Brow Pad
Material HDPE
Model H-807V
Quantity 20/Cs.
Size (Metric) 168 to 197 mm
Suspension Type 4-point Ratchet
Ventilated Ventilated
Head Style Full Brim