Learn More
3M™ H-800 Series Full Brim Vented Hard Hats with Uvicator™ Sensors
Catalog No. 17623775 Shop All 3M Products
Change view
Click to view available options
Color:
Black
Blue
Gray
Green
Hi-vis Orange
Hi-vis Yellow
Navy Blue
Orange
Pink
Red
Tan
White
Yellow
13 product options available for selection
Product selection table with 13 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
17-623-775 Black
17-623-739 White
17-623-742 Green
17-623-740 Yellow
17-623-741 Blue
17-623-743 Red
17-623-744 Orange
17-623-745 Hi-vis Orange
17-623-746 Gray
17-623-747 Hi-vis Yellow
17-623-749 Tan
17-623-748 Navy Blue
17-623-779 Pink
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 13 options available.
13 options
Catalog No. 17-623-775 Supplier 3M™ Supplier No. 7100162736
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.
With Uvicator sensor that changes color as hard hat is exposed to UV light
  • Made of HDPE
  • Features 4-point ratchet suspension with three position height adjustment allows for customized fit
  • Perforated absorbent brow pad 
  • Optional 6-point suspension strap for more comfort
  • Vents allow air circulation to keep worker cooler and more comfortable
  • Meets requirements of ANSI/ISEA Z89.1-2009 Type 1, Class C
  • Available in several colors
  • 3M Catalog# H-801V-UV, H-802V-UV, H-803V-UV, H-804V-UV, H-805V-UV, H-806V-UV, H-807V-UV, H-808V-UV, H-809V-UV, H-810V-UV, H-811V-UV, H-812V-UV, H-813V-UV
TRUSTED_SUSTAINABILITY
Specifications
Color Black
Model H-812V-UV
Quantity 20/Cs.
Head Style Full Brim